MilliporeSigma
Fragrance volatiles of developing and senescing carnation flowers.
Phytochemistry (2001-04-21)
F Schade, R L Legge, J E Thompson
ABSTRACT
Thirteen major volatiles of the carnation flower fragrance signature have been identified by GC/MS. Of these, ten, hexanal, (2E)-hexenal, 1-hexanol, 2-hexanol, 3-hexen-1-ol, nonanal, benzaldehyde, benzyl alcohol, benzyl benzoate and caryophyllene, were quantified. The steady-state levels of these ten volatiles change independently as the flowers develop and senesce, suggesting that their synthesis is developmentally regulated. In addition, the chemical composition of the fragrance signature in naturally senesced flowers proved to be very different from that for flowers that had been induced to senesce prematurely by treatment with ethylene. Thus, senescence-related changes in carnation floral scent appear not to be directly regulated by ethylene. From cellular fractionation studies, it is evident that all of the volatiles, except 2-hexanol, are present in both membranous and cytosolic compartments, suggesting that their synthesis is membrane-associated and that they subsequently partition into the cytosol in accordance with partition coefficients.