MilliporeSigma
  • Extracellular polymeric substance from Rahnella sp. LRP3 converts available Cu into Cu5(PO4)2(OH)4 in soil through biomineralization process.
Extracellular polymeric substance from Rahnella sp. LRP3 converts available Cu into Cu5(PO4)2(OH)4 in soil through biomineralization process.
Environmental pollution (Barking, Essex : 1987) (2020-05-07)
Hoaithuong Do, Chi Che, Zijun Zhao, Yuqi Wang, Mingtang Li, Xiufang Zhang, Xingmin Zhao
ABSTRACT
Soil contamination by toxic heavy metals such as copper is a serious problem. In this study, the extracellular polymeric substance (EPS) extracted from Rahnella sp. LRP3 was found with the potential of immobilizing Cu-polluted in soil. The EPS could bond to Cu (II) through functional groups (polysaccharides, amide, proteins, and carboxyl groups), which further developed into the porous sphere with a diameter of 20 μm. Besides, EPS could induce the formation of Cu5(PO4)2(OH)4 crystal by the biomineralization process. Finally, the EPS in the culture solution reduced 89.4 mg/kg of DTPA-Cu content by 78.99% in soil for 10 d under the condition of 25 °C via biomineralization. The results demonstrated that EPS produced by Rahnella sp. LRP3 will be a promising factor in the remediation of Cu contaminated soil.
Product Description
Deepwell 96 well plate, polypropylene, sterile, pack of 24 ea