MilliporeSigma
  • Respirovirus C protein inhibits activation of type I interferon receptor-associated kinases to block JAK-STAT signaling.
Respirovirus C protein inhibits activation of type I interferon receptor-associated kinases to block JAK-STAT signaling.
FEBS letters (2019-11-11)
Yoshinori Kitagawa, Mayu Yamaguchi, Miki Kohno, Madoka Sakai, Masae Itoh, Bin Gotoh
ABSTRACT
Respirovirus C protein blocks the type I interferon (IFN)-stimulated activation of the JAK-STAT pathway. It has been reported that C protein inhibits IFN-α-stimulated tyrosine phosphorylation of STATs, but the underlying mechanism is poorly understood. Here, we show that the C protein of Sendai virus (SeV), a member of the Respirovirus genus, binds to the IFN receptor subunit IFN-α/β receptor subunit (IFNAR)2 and inhibits IFN-α-stimulated tyrosine phosphorylation of the upstream receptor-associated kinases, JAK1 and TYK2. Analysis of various SeV C mutant (Cm) proteins demonstrates the importance of the inhibitory effect on receptor-associated kinase phosphorylation for blockade of JAK-STAT signaling. Furthermore, this inhibitory effect and the IFNAR2 binding capacity are observed for all the respirovirus C proteins examined. Our results suggest that respirovirus C protein inhibits activation of the receptor-associated kinases JAK1 and TYK2 possibly through interaction with IFNAR2.