MilliporeSigma
  • NCM 1921, a Mixture of Several Ingredients, Including Fatty Acids and Choline, Attenuates Atopic Dermatitis in 1-Chloro-2,4-Dinitrobenzene-Treated NC/Nga Mice.
NCM 1921, a Mixture of Several Ingredients, Including Fatty Acids and Choline, Attenuates Atopic Dermatitis in 1-Chloro-2,4-Dinitrobenzene-Treated NC/Nga Mice.
Nutrients (2020-01-16)
Young-Sil Lee, Won-Kyung Yang, Eun-Hee Jo, Seung Ho Shin, Young-Cheol Lee, Min-Cheol Park, Seung-Hyung Kim
ABSTRACT
Atopic dermatitis (AD) is a chronic inflammatory skin disease in humans. In this study, we evaluated the effects of a mixture (NCM 1921) of omega-3 butter, omega-3 beef tallow oil, omega-3 lard oil, caprylic acid, lauric acid, choline, and Fe on AD in 1-chloro-2,4-dinitrobenzene (DNCB)-treated NC/Nga mice. NCM 1921 significantly ameliorated the macroscopic and microscopic signs and reduced skin thickness and mast cell incorporation in the skin lesions of mice with DNCB-induced AD. Furthermore, it reduced serum immunoglobulin E levels; reduced the number of IgE-producing B cells, peripheral blood mononuclear cells, white blood cells, and differential white blood cells; and increased the number of lymphocytes. NCM 1921 normalized the total cell number in dorsal skin tissue, the axillary lymph node, and spleen following DNCB exposure and reduced the number of CD23+/B220+ cells in the axillary lymph node and CD3+ cells in dorsal skin tissue. Moreover, it reduced the levels of interleukin (IL)-4 and IL-13 but increased the levels of interferon-γ in anti-CD3-stimulated splenocytes. Immunohistofluorescence staining showed that NCM 1921 treatment significantly increased claudin1, filaggrin, and Sirt1 protein expressions in AD skin lesions. These results suggest that NCM 1921 could be a valuable remedy for the treatment of AD.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid