MilliporeSigma
  • Orphan nuclear receptor COUP-TFII is an oncogenic gene in renal cell carcinoma.
Orphan nuclear receptor COUP-TFII is an oncogenic gene in renal cell carcinoma.
Clinical & translational oncology : official publication of the Federation of Spanish Oncology Societies and of the National Cancer Institute of Mexico (2019-08-02)
X Fang, C-X Liu, X-R Zeng, X-M Huang, W-L Chen, Y Wang, F Ai
ABSTRACT
Chicken ovalbumin upstream promoter-transcription factor II (COUP-TFII) may be an oncogenic gene in renal cell carcinoma (RCC). However, the direct association between COUP-TFII expression and patient survival has not been investigated in patients with RCC, and the molecular oncogenesis of COUP-TFII in RCC remains unclear. The mRNA expression levels of COUP-TFII in the tumors of 283 patients with RCC were determined by RT-qPCR. The remaining 266 patients were categorized into low- and high-expression groups according to the cut off value generated by receiver operating curve (ROC) analysis. The function of COUP-TFII in RCC cells was tested by knockdown experiments in vitro. In the present study, it was revealed that the mRNA expression levels of COUP-TFII were significantly higher in tumors compared with those in adjacent non-cancerous tissues, and that the overexpression of COUP-TFII was strongly associated with poor patient survival. It was further demonstrated that knockdown of COUP-TFII suppressed proliferation, and induced apoptosis and cell cycle arrest in RCC cells in vitro. This also resulted in the activation of the mitochondria-mediated apoptosis pathway, impaired migration and invasion of RCC cells through epithelial-mesenchymal transition in vitro, and suppressed tumor growth in vivo. In addition, it was revealed that the induction of cell migration and invasion by COUP-TFII was mediated, at least in part, by integrin subunit β1. In summary, the present study indicated that COUP-TFII is an oncogenic gene in RCC, and a potential therapeutic target for the treatment of the disease.