MilliporeSigma
Metabotropic glutamate receptor activation induces astroglial swelling.
The Journal of biological chemistry (1994-09-02)
E Hansson
ABSTRACT
The activation of metabotropic glutamate receptors (mGluRs) by 1-aminocyclopentane-1,3-dicarboxylic acid (1S,3R-ACPD) or ibotenate induced a rapid cell volume increase in primary cultures of type 1 astroglial cells from the cerebral cortex of a newborn rat. These relative volume changes and parallel Ca2+ transients in single cells were examined by microspectrofluorimetry after loading the cells with fura-2/AM and varying the excitation wavelengths between the isosbestic point of the probe and its ion-sensitive wavelength. L(+)-2-Amino-4-phosphonobutyric acid (L-AP4) evoked an astroglial swelling but few or no cytosolic Ca2+ transients. No rapid swelling was observed after stimulation of ionotropic Glu receptors. The Glu-induced volume increase was unaffected by gluconate or amiloride, partially blocked by Glu-carrier blockers, and totally blocked by ketamine. The Glu- or L-AP4-induced volume increases were blocked by BaCl2 or furosemide. Tetraethylammoniumchloride-1-hydrate blocked the Glu- and 1S,3R-ACPD-induced astroglial swelling but the voltage-dependent L-, N-, or T-type Ca2+ channels were not primarily involved in the Glu-, 1S,3R-ACPD-, or L-AP4-induced swelling. mGluRs induce inositol 1,4,5-trisphosphate synthesis, intracellular Ca2+ increase, and the opening of a delayed outward K+ rectifier, and along another route they activate a Gi protein and open an inward K+ rectifier. One Na(+)-K(+)-2Cl(-)-cotransporter and a Na(+)-K(+)-ATPase is activated and so also is an electrogenic Na(+)-dependent Glu carrier. Thus, Glu-induced astroglial swelling is not only the result of the above mechanisms, but requires another, until now unidentified mechanism, probably some ketamine-sensitive K+ outflux or Na+ influx.
Sigma-Aldrich
Amiloride hydrochloride hydrate, ≥98% (HPLC), powder