MilliporeSigma
  • WiNTRLINC1/ASCL2/c-Myc Axis Characteristics of Colon Cancer with Differentiated Histology at Young Onset and Essential for Cell Viability.
WiNTRLINC1/ASCL2/c-Myc Axis Characteristics of Colon Cancer with Differentiated Histology at Young Onset and Essential for Cell Viability.
Annals of surgical oncology (2019-09-25)
Kazuko Yokota, Yoko Tanaka, Hiroki Harada, Takeshi Kaida, Syuji Nakamoto, Takafumi Soeno, Yoshiki Fujiyama, Mitsuo Yokota, Ken Kojo, Hirohisa Miura, Takahiro Yamanashi, Takeo Sato, Takatoshi Nakamura, Masahiko Watanabe, Keishi Yamashita
ABSTRACT
WiNTRLINC1 is a long non-coding RNA (lncRNA) that positively regulates the Wnt pathway via achaete-scute complex homolog 2 (ASCL2) in colorectal cancer. ASCL2 was recently reported to play a critical role in chemoresistance, however clinical relevance of the WiNTRLINC1/ASCL2 axis remains obscure in colon cancer. WiNTRLINC1/ASCL2 expression was investigated at messenger RNA (mRNA) level in 40 primary colon cancer tissues and the corresponding normal mucosa tissues, together with Wnt-related genes (c-Myc/PRL-3) and other lncRNAs (H19, HOTAIR, and MALAT1). Knock-down experiments of WiNTRLINC1 clarified its role in their expression and chemoresistance. Real-time quantitative reverse transcriptase-polymerase chain reaction confirmed definite overexpression of WiNTRLINC1 mRNA in primary colon cancer compared with the corresponding normal colon mucosa tissues (p = 0.0005), such as ASCL2, c-Myc, and PRL-3 (p < 0.0001). The four gene expression signatures were tightly associated in the center of the ASCL2 gene (r = 0.72, p < 0.0001) in clinical samples. WiNTRLINC1 was not significantly associated with prognostic factors in colon cancer and other lncRNAs, while the WiNTRLINC1/ASCL2/c-Myc signatures were unique to young-onset colon cancer with differentiated histology. On the other hand, undifferentiated histology was significantly associated with H19 expression. Knockdown of the WiNTRLINC1 gene reduced the expression of ASCL2/c-Myc, but rather augmented PRL-3 at mRNA level, and robustly affected cell viability in colon cancer cell lines. The enhanced WiNTRLINC1/ASCL2/c-Myc axis involved in Wnt pathway activation is a common pathway essential for differentiated colon tumorigenesis, especially with young onset, and may be essential for a viable phenotype of colon cancer.
Sigma-Aldrich
Nutrient Mixture F-12 Ham, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture