MilliporeSigma
  • Dysregulated Expression of Phosphorylated Epidermal Growth Factor Receptor and Phosphatase and Tensin Homologue in Canine Cutaneous Papillomas and Squamous Cell Carcinomas.
Dysregulated Expression of Phosphorylated Epidermal Growth Factor Receptor and Phosphatase and Tensin Homologue in Canine Cutaneous Papillomas and Squamous Cell Carcinomas.
Journal of comparative pathology (2020-01-21)
B L Sanz Ressel, A R Massone, C G Barbeito
ABSTRACT
The molecular mechanisms contributing to the development of cutaneous papillomas (CPs) and cutaneous squamous cell carcinomas (CSCCs) are still poorly understood, limiting the ability to identify molecular suitable targets for the development of novel therapies. Persistent activation of the phosphatidylinositol 3-kinase/Akt/mammalian target of rapamycin (PI3K/Akt/mTOR) signalling pathway is a component of epidermal carcinogenesis in dogs. The present study describes the immunohistochemical expression pattern of two key regulatory molecules involved in the PI3K/Akt/mTOR signalling pathway, phosphorylated epidermal growth factor receptor (pEGFR)Tyr1068 and phosphatase and tensin homologue (PTEN), in samples of normal canine epidermis, CP, preneoplastic epidermis and CSCC using tissue microarrays to determine whether the deregulated activity of these molecules is involved in the pathogenesis of these relevant epidermal tumours of dogs. Expression of pEGFR and PTEN was dysregulated in most samples of CP, preneoplastic epidermis and CSCC. Overexpression of pEGFR, together with decreased expression of PTEN, may facilitate the progression of some canine CPs and CSCCs by deregulation of the key cellular functions in which the PI3K/Akt/mTOR signalling pathway is involved. These findings suggest that the PI3K/Akt/mTOR signalling molecules may be potential therapeutic targets for canine patients with CP and CSCC.