MilliporeSigma
  • H19 lncRNA alters DNA methylation genome wide by regulating S-adenosylhomocysteine hydrolase.
H19 lncRNA alters DNA methylation genome wide by regulating S-adenosylhomocysteine hydrolase.
Nature communications (2015-12-22)
Jichun Zhou, Lihua Yang, Tianyu Zhong, Martin Mueller, Yi Men, Na Zhang, Juanke Xie, Karolyn Giang, Hunter Chung, Xueguang Sun, Lingeng Lu, Gordon G Carmichael, Hugh S Taylor, Yingqun Huang
ABSTRACT
DNA methylation is essential for mammalian development and physiology. Here we report that the developmentally regulated H19 lncRNA binds to and inhibits S-adenosylhomocysteine hydrolase (SAHH), the only mammalian enzyme capable of hydrolysing S-adenosylhomocysteine (SAH). SAH is a potent feedback inhibitor of S-adenosylmethionine (SAM)-dependent methyltransferases that methylate diverse cellular components, including DNA, RNA, proteins, lipids and neurotransmitters. We show that H19 knockdown activates SAHH, leading to increased DNMT3B-mediated methylation of an lncRNA-encoding gene Nctc1 within the Igf2-H19-Nctc1 locus. Genome-wide methylation profiling reveals methylation changes at numerous gene loci consistent with SAHH modulation by H19. Our results uncover an unanticipated regulatory circuit involving broad epigenetic alterations by a single abundantly expressed lncRNA that may underlie gene methylation dynamics of development and diseases and suggest that this mode of regulation may extend to other cellular components.
Sigma-Aldrich
IgG, Mouse