MilliporeSigma
  • Nuclear translocation of fibroblast growth factor-2 (FGF2) is regulated by Karyopherin-β2 and Ran GTPase in human glioblastoma cells.
Nuclear translocation of fibroblast growth factor-2 (FGF2) is regulated by Karyopherin-β2 and Ran GTPase in human glioblastoma cells.
Oncotarget (2015-06-10)
Feng Wang, Lijun Yang, Lin Shi, Qian Li, Gengshen Zhang, Jianliang Wu, Jun Zheng, Baohua Jiao
ABSTRACT
Human glioblastoma multiforme (GBM) is the most malignant tumor of the central nervous system (CNS). Fibroblast growth factor-2 (FGF2) belongs to the FGF superfamily and functions as a potential oncoprotein in GBM. FGF2 has low molecular weight (18K) and high molecular weight (HMW) isoforms. Nuclear accumulation of HMW-FGF2 strongly promotes glioblastoma cell proliferation, yet mechanism governing such cellular distribution remains unexplored. We investigated the mechanisms regulating FGF2 cellular localization in T98G human brain glioblastoma cells. We found HMW-FGF2, but not 18K-FGF2, is primarily located in the nucleus and interacts with nuclear transport protein Karyopherin-β2/Transportin (Kapβ2). SiRNA-directed Kapβ2 knockdown significantly reduced HMW-FGF2's nuclear translocation. Moreover, inhibiting Ran GTPase activity also resulted in decreased HMW-FGF2 nuclear accumulation. Proliferation of T98G cells is greatly enhanced with transfections HMW-FGF2. Decreased PTEN expression and activated Akt signaling were observed upon HMW-FGF2 overexpression and might mediate pro-survival effect of FGF2. Interestingly, addition of nuclear localization signal (NLS) to 18K-FGF2 forced its nuclear import and dramatically increased cell proliferation and Akt activation. These findings demonstrated for the first time the molecular mechanisms for FGF2's nuclear import, which promotes GBM cell proliferation and survival, providing novel insights to the development of GBM treatments.
Sigma-Aldrich
Anti-phospho-Akt (Ser473) Antibody, clone 6F5, clone 6F5, from mouse
Sigma-Aldrich
Anti-PTEN Antibody, clone 6H2.1, clone 6H2.1, Upstate®, from mouse