MilliporeSigma
  • Expression, Function, and Prognostic Value of MAGE-D4 Protein in Esophageal Squamous Cell Carcinoma.
Expression, Function, and Prognostic Value of MAGE-D4 Protein in Esophageal Squamous Cell Carcinoma.
Anticancer research (2019-11-11)
Yasuo Uno, Mitsuro Kanda, Yusuke Sato, Dai Shimizu, Shinichi Umeda, Norifumi Hattori, Masamichi Hayashi, Chie Tanaka, Daisuke Kobayashi, Suguru Yamada, Goro Nakayama, Satoru Motoyama, Masahiko Koike, Yasuhiro Kodera
ABSTRACT
We previously reported that expression of melanoma-associated antigen (MAGE)-D4 mRNA was a prognostic factor for esophageal squamous cell carcinoma (ESCC). The aim of this study was to validate the expression of MAGE-D4 in two additional patient cohorts, and to investigate its biological functions. The role of MAGE-D4 in cell proliferation, adhesion, and migration was determined by gene knockdown experiments in the KYSE590 cell line. MAGE-D4 protein expression was analyzed in ESCC tissues by immunohistochemistry. A second validation cohort consisted of an ESCC mRNA dataset from The Cancer Genome Atlas. Knockdown of MAGE-D4 significantly decreased cell proliferation and migration. Expression of MAGE-D4 protein was significantly associated with disease-free survival. In the second validation cohort, high MAGE-D4 mRNA expression was associated with significantly shorter overall survival and disease-free survival. MAGE-D4 plays an important role in the malignant behavior of ESCC. MAGE-D4 was validated as a prognostic indicator in two independent ESCC patient cohorts.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Anti-MAGED4B antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution