MilliporeSigma
  • Von Economo neurons are part of a larger neuronal population that are selectively vulnerable in C9orf72 frontotemporal dementia.
Von Economo neurons are part of a larger neuronal population that are selectively vulnerable in C9orf72 frontotemporal dementia.
Neuropathology and applied neurobiology (2019-05-09)
P Gami-Patel, I van Dijken, J C van Swieten, Y A L Pijnenburg, A J M Rozemuller, J J M Hoozemans, A A Dijkstra
ABSTRACT
The behavioural variant of frontotemporal dementia with a C9orf72 expansion (C9-bvFTD) is characterised by early changes in social-emotional cognition that are linked to the loss of von Economo neurons (VENs). Together with a subset of neighbouring pyramidal neurons, VENs express the GABA receptor subunit theta (GABRQ). It is not known if the selective vulnerability of VENs in C9-bvFTD also includes this GABRQ-expressing population. We quantified VENs and GABRQ immunopositive neurons in the anterior cingulate cortex (ACC) in C9-bvFTD (n = 16), controls (n = 12) and Alzheimer's disease (AD) (n = 7). Second, we assessed VENs and GABRQ-expressing populations in relation to the clinicopathological profiles. We found the number of VENs and GABRQ-expressing neurons and their ratio over the total layer 5 neuronal population was lower in C9-bvFTD compared to control and AD. C9-bvFTD donors with underlying TDP43 type A pathology in the ACC showed the highest loss of GABRQ-expressing neurons. C9-bvFTD donors that did not present with motor neuron disease (MND) symptoms in the first half of their disease course showed a prominent loss of GABRQ-expressing neurons compared to controls. C9-bvFTD donors with no symptoms of psychosis showed a higher loss compared to controls. Across all donors, the number of VENs correlated strongly with the number of GABRQ-expressing neurons. We show that VENs, together with GABRQ-expressing neurons, are selectively vulnerable in C9-bvFTD but are both spared in AD. This suggests they are related and that this GABRQ-expressing population of VENs and pyramidal neurons, is a key modulator of social-emotional functioning.
Sigma-Aldrich
Anti-GABRE antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution