MilliporeSigma
  • Network-Specific Synchronization of Electrical Slow-Wave Oscillations Regulates Sleep Drive in Drosophila.
Network-Specific Synchronization of Electrical Slow-Wave Oscillations Regulates Sleep Drive in Drosophila.
Current biology : CB (2019-10-22)
Davide Raccuglia, Sheng Huang, Anatoli Ender, M-Marcel Heim, Desiree Laber, Raquel Suárez-Grimalt, Agustin Liotta, Stephan J Sigrist, Jörg R P Geiger, David Owald
ABSTRACT
Slow-wave rhythms characteristic of deep sleep oscillate in the delta band (0.5-4 Hz) and can be found across various brain regions in vertebrates. Across phyla, however, an understanding of the mechanisms underlying oscillations and how these link to behavior remains limited. Here, we discover compound delta oscillations in the sleep-regulating R5 network of Drosophila. We find that the power of these slow-wave oscillations increases with sleep need and is subject to diurnal variation. Optical multi-unit voltage recordings reveal that single R5 neurons get synchronized by activating circadian input pathways. We show that this synchronization depends on NMDA receptor (NMDAR) coincidence detector function, and that an interplay of cholinergic and glutamatergic inputs regulates oscillatory frequency. Genetically targeting the coincidence detector function of NMDARs in R5, and thus the uncovered mechanism underlying synchronization, abolished network-specific compound slow-wave oscillations. It also disrupted sleep and facilitated light-induced wakening, establishing a role for slow-wave oscillations in regulating sleep and sensory gating. We therefore propose that the synchronization-based increase in oscillatory power likely represents an evolutionarily conserved, potentially "optimal," strategy for constructing sleep-regulating sensory gates.
Product Description
Supelco
Calcium ionophore I, Selectophore, function tested, ≥99.0%
Sigma-Aldrich
D(−)-2-Amino-5-phosphonopentanoic acid, NMDA receptor antagonist
Sigma-Aldrich
all trans-Retinal, powder, ≥98%