MilliporeSigma
  • Seasonal variation, source apportionment and source attributed health risk of fine carbonaceous aerosols over National Capital Region, India.
Seasonal variation, source apportionment and source attributed health risk of fine carbonaceous aerosols over National Capital Region, India.
Chemosphere (2019-09-25)
Shivani, Ranu Gadi, Sudhir Kumar Sharma, Tuhin Kumar Mandal
ABSTRACT
Deteriorating air quality with high levels of fine particulate matter (PM2.5) over National Capital Region (NCR) of India is one of the serious environmental and scientific issues. In this paper, PM2.5 samples were collected for 24 h twice or thrice a week during December 2016-December 2017 at three sites [Delhi (IG), Modinagar (MN) and Mahendragarh (HR)] over NCR to analyse the carbonaceous aerosols. Source apportionment of PM2.5 was attempted using Principal Component analysis (PCA) and Positive Matrix Factorization (PMF) based on the analysed carbonaceous fractions [Organic carbon, Elemental carbon, Secondary organic carbon (SOC)]. Organic compounds: alkanes, hopanes, steranes, polycyclic aromatic hydrocarbons (PAHs), phthalates, levoglucosan and n-alkanoic acids were analysed to distinguish the emission sources. Total Carbonaceous Aerosols (TCA) contributed significantly (∼26%) to PM2.5 which revealed their importance in source apportionment. Estimated SOC contributed 43.2%, 42.2% and 58.2% to OC and 5.4%, 5.3% and 7.8% to PM2.5 at IG, MN and HR sites respectively. PCA and PMF apportion five emission sources i.e., vehicular emissions (34.6%), biomass burning (26.8%), cooking emissions (15.7%), plastic and waste burning (13.5%) and secondary organic carbon (9.5%) for PM2.5. Source attributed health risk has also been calculated in terms of Lung cancer risk (LCR) associated with PAHs exposure and concluded that vehicular emissions (40.3%), biomass burning (38.1%), secondary organic carbon (12.8%) contributed higher to LCR (503.2 × 10-5; ∼503 cases in 1,00,000). Health risk assessment combined with source apportionment inferences signifies the immediate implementation of emissions reduction strategies with special target on transport sector and biomass burning over the NCR of India.
Product Description
Supelco
Polynuclear aromatic hydrocarbons 16 solution, analytical standard, in acetonitrile
Supelco
C8-C40 Alkanes Calibration Standard, in dichloromethane (varied)