MilliporeSigma
  • Synthesis of New Fused Heterocyclic 2-Quinolones and 3-Alkanonyl-4-Hydroxy-2-Quinolones.
Synthesis of New Fused Heterocyclic 2-Quinolones and 3-Alkanonyl-4-Hydroxy-2-Quinolones.
Molecules (Basel, Switzerland) (2019-10-24)
Ashraf A Aly, Alaa A Hassan, Nasr K Mohamed, Lamiaa E Abd El-Haleem, Stefan Bräse, Mika Polamo, Martin Nieger, Alan B Brown
ABSTRACT
Herein, we report the synthesis of 5,12-dihydropyrazino[2,3-c:5,6-c']difuro[2,3-c:4,5-c']-diquinoline-6,14(5H,12H)diones, 2-(4-hydroxy-2-oxo-1,2-dihydroquinolin-3-yl)-1,4-diphenyl- butane-1,4-diones and 4-(benzo-[d]oxazol-2-yl)-3-hydroxy-1H-[4,5]oxazolo[3,2-a]pyridine-1-one. The new candidates were synthesized and identified by different spectroscopic techniques, and X-ray crystallography.
Sigma-Aldrich
trans-1,4-Diphenyl-2-butene-1,4-dione, 96%