MilliporeSigma
Preserve Your Books through the Smell.
ACS sensors (2019-10-28)
Marta I S Veríssimo, João A B P Oliveira, Dmitry V Evtuguin, M Teresa S R Gomes
ABSTRACT
The identification of paper composition, pH, early signs of paper degradation, and emitting volatile organic compounds (VOCs) are mandatory for effective preventive conservation of paper/books. Sampling restrictions in the analysis of cultural heritage materials limit the choice of appropriate analytical methods. Solvent-free analytical techniques with nondestructive sampling are needed. Addressing this challenge, an electronic nose based on six-coated piezoelectric quartz crystals was assembled and used to analyze VOCs emitted from books. Careful selection of sensor coatings and cluster analysis allowed us to achieve a clear distinction between cotton/linen rag and wood pulp paper, and among the letter group, the discrimination between papers manufactured from alkaline and acidic stocks. This six-element sensor array was therefore able to replace destructive tests as the ones ordinarily used for paper pH measurements. The same electronic nose was able to separate aged pale-yellow paper, a visible initial sign of paper degradation, from well-preserved still white papers, even when made from the same raw material. One of the used sensors detected furfural, often seen as a marker of cellulose degradation, at lower levels than the detection limit found in the literature with gas chromatography-mass spectrometry, a much more complex bulky and expensive instrument.
Sigma-Aldrich
Manganese(II) phthalocyanine
Sigma-Aldrich
N,N,N′,N′-Tetrakis(2-hydroxyethyl)ethylenediamine, technical
Sigma-Aldrich
N-Ethylethylenediamine, 98%