MilliporeSigma
  • A vaccinology Approach to the Identification and Characterization of Dermanyssus Gallinae Candidate Protective Antigens for the Control of Poultry Red Mite Infestations.
A vaccinology Approach to the Identification and Characterization of Dermanyssus Gallinae Candidate Protective Antigens for the Control of Poultry Red Mite Infestations.
Vaccines (2019-11-24)
José Francisco Lima-Barbero, Marinela Contreras, Lourdes Mateos-Hernández, Francisco Manuel Mata-Lorenzo, Roxana Triguero-Ocaña, Olivier Sparagano, Robert D Finn, Christina Strube, Daniel R G Price, Francesca Nunn, Kathryn Bartley, Ursula Höfle, Mariana Boadella, Alasdair J Nisbet, José de la Fuente, Margarita Villar
ABSTRACT
The poultry red mite (PRM), Dermanyssus gallinae, is a hematophagous ectoparasite considered as the major pest in the egg-laying industry. Its pesticide-based control is only partially successful and requires the development of new control interventions such as vaccines. In this study, we follow a vaccinology approach to identify PRM candidate protective antigens. Based on proteomic data from fed and unfed nymph and adult mites, we selected a novel PRM protein, calumenin (Deg-CALU), which is tested as a vaccine candidate on an on-hen trial. Rhipicephalusmicroplus Subolesin (Rhm-SUB) was chosen as a positive control. Deg-CALU and Rhm-SUB reduced the mite oviposition by 35 and 44%, respectively. These results support Deg-CALU and Rhm-SUB as candidate protective antigens for the PRM control.
Product Description
AeraSeal film, sterile