MilliporeSigma
  • Genome-wide Profiling of Histone Lysine Butyrylation Reveals its Role in the Positive Regulation of Gene Transcription in Rice.
Genome-wide Profiling of Histone Lysine Butyrylation Reveals its Role in the Positive Regulation of Gene Transcription in Rice.
Rice (New York, N.Y.) (2019-11-30)
Shuai Liu, Guanqing Liu, Peifeng Cheng, Chao Xue, Yong Zhou, Xu Chen, Lu Ye, Zhongying Qiao, Tao Zhang, Zhiyun Gong
ABSTRACT
Histone modifications play important roles in growth and development of rice (Oryza sativa L.). Lysine butyrylation (Kbu) with a four-carbon chain is a newly-discovered histone acylation modification in rice. In this study, we performed chromatin immunoprecipitation sequencing (ChIP-seq) analyses, the result showed that major enrichment of histone Kbu located in genebody regions of rice genome, especially in exons. The enrichment level of Kbu histone modification is positively correlated with gene expression. Furthermore, we compared Kbu with DNase-seq and other histone modifications in rice. We found that 60.06% Kub enriched region co-located with DHSs in intergenic regions. The similar profiles were detected among Kbu and several acetylation modifications such as H3K4ac, H3K9ac, and H3K23ac, indicating that Kbu modification is an active signal of transcription. Genes with both histone Kbu and one other acetylation also had significantly increased expression compared with genes with only one acetylation. Gene Ontology (GO) enrichment analysis revealed that these genes with histone Kbu can regulate multiple metabolic process in different rice varieties. Our study showed that the lysine butyrylation modificaiton may promote gene expression as histone acetylation and will provide resources for futher studies on histone Kbu and other epigenetic modifications in plants.
Product Description
rProtein A Sepharose Fast Flow, Cytiva 17-1279-01, pack of 5 mL