MilliporeSigma
  • Continuous UiO-66-Type Metal-Organic Framework Thin Film on Polymeric Support for Organic Solvent Nanofiltration.
Continuous UiO-66-Type Metal-Organic Framework Thin Film on Polymeric Support for Organic Solvent Nanofiltration.
ACS applied materials & interfaces (2019-11-14)
Dangchen Ma, Gang Han, Zhuo Fan Gao, Shing Bor Chen
ABSTRACT
For the first time, continuous polycrystalline UiO-66-NH2 thin film supported by a cross-linked Matrimid substrate was successfully fabricated via in situ solvothermal synthesis at room temperature for organic solvent nanofiltration. The integrated structure of the formed UiO-66-NH2 selective layer was inferred by various characterizations including X-ray diffraction, field-emission scanning electron microscopy, energy-dispersive X-ray, Fourier transform infrared spectroscopy, and X-ray photoelectron spectroscopy. We have demonstrated that pretreatment of the substrate by an organic ligand, the number of solvothermal synthesis cycles, and the reaction time play important roles in MOF film formation. The newly developed UiO-66-NH2 membrane possesses high surface hydrophobicity and mean pore size of 0.89 nm in diameter. It shows an exceptional rejection of 96.33% to Rose Bengal with moderate ethanol permeance of 0.88 L m-2 h-1 bar-1. Benefiting from the extraordinary chemical stability of Zr-MOF crystals, the UiO-66-NH2 membrane shows excellent stability in different solvents, implying their great potential for real applications. This work provides useful insights into the fabrication of continuous UiO-66-type MOF membranes on polymeric substrates, which are very promising in practical separations involving organic solvents.
Sigma-Aldrich
2-Aminoterephthalic acid, 99%