MilliporeSigma
  • Investigation of Pyrophosphates KYP2O7Co-Doped with Lanthanide Ions Useful for Theranostics.
Investigation of Pyrophosphates KYP2O7Co-Doped with Lanthanide Ions Useful for Theranostics.
Nanomaterials (Basel, Switzerland) (2019-11-14)
Adam Watras, Marta Wujczyk, Michael Roecken, Katarzyna Kucharczyk, Krzysztof Marycz, Rafal J Wiglusz
ABSTRACT
Diphosphate compounds (KYP2O7) co-doped with Yb3+ and Er3+ ions were obtained by one step urea assisted combustion synthesis. The experimental parameters of synthesis were optimized using an experimental design approach related to co-dopants concentration and heattreatment as well as annealing time. The obtained materials were studied with theinitial requirements showing appropriate morphological (X-Ray Diffraction (XRD), Scanning Electron Microscopy (SEM)) and spectroscopic properties (emission, luminescence kinetics). Moreover, the effect of Er3+ and Yb3+ ions doped KYP2O7 on morphology, proliferative and metabolic activity and apoptosis in MC3T3-E1 osteoblast cell line and 4B12osteoclasts cell line was investigated. Furthermore, the expression of the common pro-osteogenic markers in MC3T3-E1 osteoblast as well as osteoclastogenesis related markers in 4B12 osteoclasts was evaluated. The extensive in vitro studies showed that KYP2O7 doped with 1 mol% Er3+ and 20 mol% Yb3+ ions positively affected the MC3T3-E1 and 4B12 cells activity without triggering their apoptosis. Moreover, it was shown that an activation of mTOR and Pi3k signaling pathways with 1 mol% Er3+, 20 mol% Yb3+: KYP2O7 can promote the MC3T3-E1 cells expression of late osteogenic markers including RUNX and BMP-2. The obtained data shed a promising light for KYP2O7 doped with Er3+ and Yb3+ ions as a potential factors improving bone fracture healing as well as in bioimaging (so-called in theranostics).
Sigma-Aldrich
Minimum Essential Medium Eagle, Alpha Modification, With sodium bicarbonate and L-glutamine, without ribonucleosides and deoxyribonucleosides, liquid, sterile-filtered, suitable for cell culture