MilliporeSigma
  • Enhanced Triacylglycerol Content and Gene Expression for Triacylglycerol Metabolism, Acyl-Ceramide Synthesis, and Corneocyte Lipid Formation in the Epidermis of Borage Oil Fed Guinea Pigs.
Enhanced Triacylglycerol Content and Gene Expression for Triacylglycerol Metabolism, Acyl-Ceramide Synthesis, and Corneocyte Lipid Formation in the Epidermis of Borage Oil Fed Guinea Pigs.
Nutrients (2019-11-23)
Ju-Young Lee, Kwang-Hyeon Liu, Yunhi Cho, Kun-Pyo Kim
ABSTRACT
Triacylglycerol (TAG) metabolism is related to the acyl-ceramide (Cer) synthesis and corneocyte lipid envelope (CLE) formation involved in maintaining the epidermal barrier. Prompted by the recovery of a disrupted epidermal barrier with dietary borage oil (BO: 40.9% linoleic acid (LNA) and 24.0% γ-linolenic acid (GLA)) in essential fatty acid (EFA) deficiency, lipidomic and transcriptome analyses and subsequent quantitative RT-PCR were performed to determine the effects of borage oil (BO) on TAG content and species, and the gene expression related to overall lipid metabolism. Dietary BO for 2 weeks in EFA-deficient guinea pigs increased the total TAG content, including the TAG species esterified LNA, GLA, and their C20 metabolized fatty acids. Moreover, the expression levels of genes in the monoacylglycerol and glycerol-3-phosphate pathways, two major pathways of TAG synthesis, increased, along with those of TAG lipase, acyl-Cer synthesis, and CLE formation. Dietary BO enhanced TAG content, the gene expression of TAG metabolism, acyl-Cer synthesis, and CLE formation.
Sigma-Aldrich
Glyceryl trioleate, ~65%
Triolein, European Pharmacopoeia (EP) Reference Standard