MilliporeSigma
  • Effects of particle size and lipid form of corn on energy and nutrient digestibility in diets for growing pigs.
Effects of particle size and lipid form of corn on energy and nutrient digestibility in diets for growing pigs.
Asian-Australasian journal of animal sciences (2019-09-05)
Zhiqian Lyu, Lu Wang, Yifan Wu, Chengfei Huang
ABSTRACT
Two experiments were conducted to evaluate the effects of corn particle size and lipid form on the apparent total tract digestibility (ATTD) of energy and nutrients in diets for growing pigs. In Exp. 1, thirty barrows (initial body weight [BW], 53.1±3.9 kg) were allotted to 1 of 5 diets formulated with 96.9% corn ground to 441, 543, 618, 659, and 768 μm, respectively. In Exp. 2, thirty-six barrows (initial BW, 54.7±3.6 kg) were allotted to 1 of 6 diets formulated by including 2% or 15% corn germ (CG 2 or CG 15), 1% or 6% corn oil (CO 1 or CO 6), 1% CO+2% corn germ meal (CO 1+CGM 2), or 6% CO+15% corn germ meal (CO 6+CGM 15), respectively. The ATTD of gross energy (GE) and the digestible energy (DE) in diet and corn grain linearly decreased as the corn particle size increased (p<0.05) from 441 to 768 μm. Particle size had a quadratic effect (p<0.05) on the ATTD of neutral detergent fiber and acid detergent fiber in diets, and which firstly increased and then decreased as the corn particle size increased from 441 to 618 μm and 618 to 768 μm, respectively. The ATTD of GE, ether extract (EE), and the DE in CO 1 diet and CO 6 diet was greater (p<0.05) than that in CG 2 diet and CG 15 diet, respectively. The ATTD of EE in CO 6 diet and CO 6+CGM 15 diet was greater (p<0.05) than that in CO 1 diet and CO 1+CGM 2 diet. Less than 618 μm was recommended for corn particle size in growing pig's diet and extracted lipid had greater digestibility than the intact lipid in corn. Higher concentration of extracted CO had greater digestibility of EE compared with lower concentrations of CO diet.
Product Description
Supelco
Starch (GO/P) Assay Kit, sufficient for 20 assays