MilliporeSigma
  • HIF-1α/BNIP3 signaling pathway-induced-autophagy plays protective role during myocardial ischemia-reperfusion injury.
HIF-1α/BNIP3 signaling pathway-induced-autophagy plays protective role during myocardial ischemia-reperfusion injury.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2019-10-08)
Yanan Zhang, Dawei Liu, Haijuan Hu, Puqiang Zhang, Ruiqin Xie, Wei Cui
ABSTRACT
The study was established to inquire into the protective effect of the HIF-1α (Hypoxia-inducible factor-1α)/ BNIP3(Bcl-2/adenovirus E1B 19-kDa interacting protein) signal path-induced-autophagy during myocardial ischemia/ reperfusion (I/R) and oxygen-glucose deprivation/recovery (OGD/R) injury in heart-derived H9C2 cells as well as its potential underlying mechanism. Immediate myocardial I/R in SD (Spraque Dawley) rats and cytotoxicity of OGD/R injury on H9C2 cells with and without inhibitors or agonists of HIF-1α and BNIP3 were evaluated. Expression of mitochondrial autophagic protein were detected by Western blot and immunofluorescence. And the mitochondrial autophagosome were detected using Transmission Electron Microscope (TEM). I/R and OGD/R injury increased the expression level of HIF-1α, activated the downstream BNIP3 and subsequently triggered mitochondria-dependent autophagy. Up-regulation the expression of HIF-1α and BNIP3 may promote the cardiac myocytes of SD rats of I/R injure and OGD/R injury-induced autophagy of H9C2 cells. Moreover, down-regulation the expression of HIF-1α or BNIP3-siRNA decreased H9C2 cells autophagy under OGD/R injury. Together, our studies indicated that HIF-1α synchronization regulate BNIP3 during OGD/R injury-induced autophagy in H9C2 cells, though BNIP3-induced autophagy acting as a survival mechanism.
Sigma-Aldrich
MISSION® esiRNA, targeting human BNIP3