MilliporeSigma
  • Current Effects of Cyanobacteria Toxin in Water Sources and Containers in the Hartbeespoort Dam Area, South Africa.
Current Effects of Cyanobacteria Toxin in Water Sources and Containers in the Hartbeespoort Dam Area, South Africa.
International journal of environmental research and public health (2019-11-27)
Matodzi Michael Mokoena, Murembiwa Stanley Mukhola
ABSTRACT
The study investigated the effects of cyanobacteria toxins such as microcystins in water sources and water stored in containers during its blooming and decaying seasons. Samples from water sources and containers near the Hartbeespoort Dam in South Africa were analysed using a microcystin ELIZA test kit. Microcystins were present in water sources used by the community, with an average of 4.3 μg/L in communal tap water and 4.8 μg/L in the water stored in tanks. The concentration of microcystins was lower in groundwater in the decaying season (0.38 μg/L) than in the blooming season (1.4 μg/L). Although microcystins were present in the storage containers, the average levels in all water samples were below the acceptable limit of 1 μg/L. The present study confirmed the presence of microcystins in the water storage containers. Therefore, it is suggested that water used for drinking from community water sources should be treated before storage to eliminate microcystins.
Product Description
BRAND® micro pipette controller, ejects disposable micropipettes, without touching them