MilliporeSigma
  • Photochemical regulation of the activity of an endonuclease BamHI using an azobenzene moiety incorporated site-selectively into the dimer interface.
Photochemical regulation of the activity of an endonuclease BamHI using an azobenzene moiety incorporated site-selectively into the dimer interface.
Chemical communications (Cambridge, England) (2004-10-30)
Koji Nakayama, Masayuki Endo, Tetsuro Majima
ABSTRACT
Endonuclease BamHI mutants having an azophenylalanine residue in the dimer interface (azoAla-BamHI) were synthesized; while the activity was almost suppressed using trans-azoAla-BamHI, the cis-isomer generated with photoirradiation recovered its intrinsic activity.
Sigma-Aldrich
Phenylalanine-4′-azobenzene HCl, ≥95%