MilliporeSigma
  • Functionalization of GlucoPyranosides at position 5 by 1,5 C-H insertion of Rh(II)-Carbenes: Dramatic influence of the anomeric configuration.
Functionalization of GlucoPyranosides at position 5 by 1,5 C-H insertion of Rh(II)-Carbenes: Dramatic influence of the anomeric configuration.
Carbohydrate research (2019-11-07)
Jana Hammoud, Antoine Joosten, Thomas Lecourt
ABSTRACT
Herein, a C-H bond functionalization approach is reported as a new route towards non-natural glycosides having a quaternary position 5. The development of this transformation furthermore reveals that insertion of Rh(II)-carbenes into the C5-H bond is controlled by remote stereoelectronic effects induced by the axial or equatorial orientation of the aglycone.
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, ZerO2®, 99.8%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%