MilliporeSigma
  • Comparison of citrus pectin and apple pectin in conjugation with soy protein isolate (SPI) under controlled dry-heating conditions.
Comparison of citrus pectin and apple pectin in conjugation with soy protein isolate (SPI) under controlled dry-heating conditions.
Food chemistry (2019-11-05)
Xiaobin Ma, Weijun Chen, Tianyi Yan, Danli Wang, Furong Hou, Song Miao, Donghong Liu
ABSTRACT
In this study, we selected two most commonly-available commercial pectin, i.e. citrus pectin and apple pectin as the grafting polysaccharides to prepare soy protein isolate-pectin conjugates. Despite the similar degrees of methoxylation and acetylation for two pectin samples, apple pectin showed much more complex structures compared to citrus pectin, with a 2.20-fold higher molecular weight and large numbers of side chains. The conjugates were prepared under controlled dry-heating conditions and achieved the degree of graft of 25.00% and 21.85% for citrus and apple pectin, respectively. Formation of the conjugates was further confirmed by SDS-PAGE gel electrophoresis and IR spectra. Attributed to the strong steric-hindrance effect of pectin, the fluorescence intensity and surface hydrophobicity of the soy protein isolate were significantly decreased after Maillard reaction. However, both solubility and emulsifying properties of the conjugates were significantly improved. Results indicated that both pectin samples played favorable roles in protein modification.
Sigma-Aldrich
Pectin from apple
Sigma-Aldrich
Pectin from citrus peel, Galacturonic acid ≥74.0 % (dried basis)
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG