MilliporeSigma
  • Dimeric benzoboroxoles for targeted activity against Mycobacterium tuberculosis.
Dimeric benzoboroxoles for targeted activity against Mycobacterium tuberculosis.
Organic & biomolecular chemistry (2019-10-30)
Collette S Guy, Kathryn Murray, Matthew I Gibson, Elizabeth Fullam
ABSTRACT
Dimeric benzoboroxoles that are covalently linked by a short scaffold enhance selective anti-tubercular activity. These multimeric benzoboroxole compounds are capable of engaging the specific extracellular Mycobacterium tuberculosis glycans, do not lead to the evolution of resistance and bypass the need to cross the impermeable mycobacterial cell envelope barrier.
Sigma-Aldrich
5-(Biotinamido)pentylamine TFA Salt, ≥95%