MilliporeSigma
  • The long noncoding RNA HOTAIR activates the Hippo pathway by directly binding to SAV1 in renal cell carcinoma.
The long noncoding RNA HOTAIR activates the Hippo pathway by directly binding to SAV1 in renal cell carcinoma.
Oncotarget (2017-09-25)
Guanghui Hu, Binbin Dong, Jingwei Zhang, Wei Zhai, Tiancheng Xie, Bisheng Huang, Chi Huang, Xudong Yao, Junhua Zheng, Jianping Che, Yun-Fei Xu
ABSTRACT
The long noncoding RNA HOTAIR promotes the development and progression of several tumors. Here, the clinical significance and role of HOTAIR in renal cell carcinoma (RCC) tumorigenesis were explored. The results showed that increased expression of HOTAIR predicted a poor prognosis of RCC after surgery. HOTAIR promoted RCC cell proliferation and growth in vitro and in vivo. The expressions of HOTAIR and Salvador homolog 1 (SAV1) were inversely correlated in clinical RCC samples. HOTAIR downregulated SAV1 by directly binding to the SAV1 protein and enhanced histone H3K27 methylation. Loss of function of SAV1 activated the Hippo pathway. HOTAIR could be a potential therapeutic target in RCC.
Sigma-Aldrich
GSK503, ≥98% (HPLC)