MilliporeSigma
  • Alcohol Withdrawal and Proopiomelanocortin Neuropeptides in an Animal Model of Alcohol Dependence.
Alcohol Withdrawal and Proopiomelanocortin Neuropeptides in an Animal Model of Alcohol Dependence.
Neuropsychobiology (2019-05-23)
Lars Hendrik Müschen, Mathias Rhein, Viktoria Hoppe, Nadine John, Kerstin Schwabe, Helge Frieling, Stefan Bleich, Marc André Nicolas Muschler
ABSTRACT
Alcohol is one of the leading threats to health worldwide. Craving for alcohol makes abstinence a difficult challenge by maintaining alcohol dependence. Many studies suppose the hypothalamic-pituitary-adrenal axis, especially the proopiomelanocortin (POMC)-derived neuropeptides, to mediate craving during withdrawal in alcohol dependence. Evidence is available that the two POMC proteins, α-melanocyte-stimulating hormone (α-MSH) and β-endorphin (β-END) are altered by alcohol consumption and influence alcohol consumption, respectively. We investigated the dynamics of α-MSH and β-END during alcohol withdrawal and the influence of intraperitoneal administration of either α-MSH or β-END in an established rodent model (Wistar rats) for alcohol dependence. After long-term alcohol self-administration over 12 months and repeated deprivation periods for 3 days, we found a significant decrease in α-MSH levels during withdrawal in rodents (p = 0.006) compared to controls, while β-END levels remained unchanged. Treatment with intraperitoneally administered α-MSH and β-END did not affect alcohol drinking behavior after deprivation. We demonstrate the effects of alcohol deprivation on α-MSH in alcohol-dependent rodents, which appear to mimic α-MSH alteration found after fasting periods during appetite regulation. Therefore, low α-MSH levels are a possible indicator for craving in alcohol-dependent individuals and hence would be a potential target for anti-craving treatment.
Sigma-Aldrich
β-Endorphin rat, ≥97% (HPLC)
Sigma-Aldrich
α-Melanocyte stimulating hormone, ≥97% (HPLC)
Sigma-Aldrich
Sodium chloride, puriss. p.a., ≥99.5% (AT)