MilliporeSigma
  • Therapeutic preferability of gemcitabine for ARID1A-deficient ovarian clear cell carcinoma.
Therapeutic preferability of gemcitabine for ARID1A-deficient ovarian clear cell carcinoma.
Gynecologic oncology (2019-10-13)
Takafumi Kuroda, Hideaki Ogiwara, Mariko Sasaki, Kazuaki Takahashi, Hiroshi Yoshida, Takako Kiyokawa, Kazuki Sudo, Kenji Tamura, Tomoyasu Kato, Aikou Okamoto, Takashi Kohno
ABSTRACT
Ovarian clear cell carcinoma (OCCC) is often resistant to conventional, standard chemotherapy using cytotoxic drugs. OCCC harbors a unique genomic feature of frequent (approximately 50%) ARID1A deficiency. The present study was performed to investigate standard chemotherapeutic options suitable for ARID1A-deficient OCCC patients. Drugs with selective toxicity to ARID1A-deficient OCCC cells were identified among six cytotoxic drugs used in standard chemotherapy for OCCC by employing multiple ARID1A-knockout cell lines and an OCCC cell line panel. Anti-tumor effects of drug treatment were assessed using a xenograft model. To obtain proof of concept in patients, seven OCCC patients who received single-agent therapy with gemcitabine were identified in a retrospective cohort of 149 OCCC patients. Patient samples and cases were analyzed for association between therapeutic response and ARID1A deficiency. ARID1A-knockout and ARID1A-deficient OCCC cells had selective sensitivity to gemcitabine. IC50 values for gemcitabine of ARID1A-deficient cells were significantly lower than those of ARID1A-proficient cells (p = 0.0001). Growth of OCCC xenografts with ARID1A deficiency was inhibited by administration of gemcitabine, and gemcitabine treatment effectively induced apoptosis in ARID1A-deficient OCCC cells. Three ARID1A-deficient OCCC patients had significantly longer progression-free survival after gemcitabine treatment than four ARID1A-proficient OCCC patients (p = 0.02). An ARID1A-deficient case that was resistant to multiple cytotoxic drugs, including paclitaxel plus carboplatin in the adjuvant and etoposide plus irinotecan in the first-line treatment, exhibited a dramatic response to gemcitabine in the second-line treatment. ARID1A-deficient OCCC patients could benefit from gemcitabine treatment in clinical settings.
Sigma-Aldrich
Pemetrexed disodium heptahydrate, ≥98% (HPLC)