MilliporeSigma
  • Robust prediction of HLA class II epitopes by deep motif deconvolution of immunopeptidomes.
Robust prediction of HLA class II epitopes by deep motif deconvolution of immunopeptidomes.
Nature biotechnology (2019-10-16)
Julien Racle, Justine Michaux, Georg Alexander Rockinger, Marion Arnaud, Sara Bobisse, Chloe Chong, Philippe Guillaume, George Coukos, Alexandre Harari, Camilla Jandus, Michal Bassani-Sternberg, David Gfeller
ABSTRACT
Predictions of epitopes presented by class II human leukocyte antigen molecules (HLA-II) have limited accuracy, restricting vaccine and therapy design. Here we combined unbiased mass spectrometry with a motif deconvolution algorithm to profile and analyze a total of 99,265 unique peptides eluted from HLA-II molecules. We then trained an epitope prediction algorithm with these data and improved prediction of pathogen and tumor-associated class II neoepitopes.
Product Description
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile