MilliporeSigma
  • Water hydrogen uptake in biomolecules detected via nuclear magnetic phosphorescence.
Water hydrogen uptake in biomolecules detected via nuclear magnetic phosphorescence.
Scientific reports (2019-11-21)
Aude Sadet, Cristina Stavarache, Florin Teleanu, Paul R Vasos
ABSTRACT
We introduce a new symmetry-based method for structural investigations of areas surrounding water-exchanging hydrogens in biomolecules by liquid-state nuclear magnetic resonance spectroscopy. Native structures of peptides and proteins can be solved by NMR with fair resolution, with the notable exception of labile hydrogen sites. The reason why biomolecular structures often remain elusive around exchangeable protons is that the dynamics of their exchange with the solvent hampers the observation of their signals. The new spectroscopic method we report allows to locate water-originating hydrogens in peptides and proteins via their effect on nuclear magnetic transitions similar to electronic phosphorescence, long-lived coherences. The sign of long-lived coherences excited in coupled protons can be switched by the experimenter. The different effect of water-exchanging hydrogens on long-lived coherences with opposed signs allows to pinpoint the position of these labile hydrogen atoms in the molecular framework of peptides and proteins.
Sigma-Aldrich
Ala-Gly