- In vivo biotinylated recombinant influenza A virus hemagglutinin for use in subtype-specific serodiagnostic assays.
In vivo biotinylated recombinant influenza A virus hemagglutinin for use in subtype-specific serodiagnostic assays.
There is an urgent need for robust subtype-specific serological tests to diagnose influenza A virus infections in poultry and mammals, including humans. Such assays require reliable subtype-specific sources of soluble and authentically folded seroreactive hemagglutinin (HA), one of the integral membrane proteins that determine the serological subtype of influenza viruses. To this purpose, a bigenic pFastBacDual baculovirus transfer vector allowing efficient invivo biotinylation of soluble HA homo-oligomers expressed via the secretory pathway was developed. An Avi-Tag allowed site-specific biotinylation by a coexpressed genetically modified BirA biotin ligase retained in the endoplasmic reticulum (ER). Highly seroreactive mono-biotinylated HA of recent H5 and H7 influenza A subtypes was secreted from recombinant baculovirus infected High-Five insect cells at levels sufficient to directly load streptavidin-coated enzyme-linked immunosorbent assay (ELISA) matrices, thereby avoiding any purification steps. The recombinant antigens retained authentic antigenicity, including conformation-dependent epitopes involved in hemagglutination inhibition as detected by monoclonal antibodies. This is the first bigenic invivo biotinylation system established for use in insect cells with secretable recombinant membrane proteins biotinylated by an ER-retained variant of BirA biotin ligase. The proposed technique is expected to significantly increase flexibility in the design of subtype-specific assays, thereby expanding the power of influenzaA virus serodiagnosis.