MilliporeSigma
  • Genome-scale analysis of library sorting (GALibSo): Isolation of secretion enhancing factors for recombinant protein production in Pichia pastoris.
Genome-scale analysis of library sorting (GALibSo): Isolation of secretion enhancing factors for recombinant protein production in Pichia pastoris.
Biotechnology and bioengineering (2009-10-10)
Gerhard Stadlmayr, Katrin Benakovitsch, Brigitte Gasser, Diethard Mattanovich, Michael Sauer
ABSTRACT
A method combining fluorescence activated cell sorting (FACS) and DNA microarray assisted clone identification was developed and termed Genome-Scale Analysis of Library Sorting (GALibSo). Genes enhancing the production of secreted heterologous proteins in Pichia pastoris were identified out of a cDNA library by cell surface display and FACS. The trends of gene enrichment during consecutive FACS rounds were monitored by DNA microarrays. In a case study a P. pastoris cDNA library was co-expressed in a strain secreting the Fab fragment of a monoclonal antibody against human immunodeficiency virus type 1 as a model protein. Three genes were identified, increasing the relative expression level of the surface-displayed model protein up to 45%. While one of these genes had a positive effect on three out of four tested proteins, the product specific effect of the other two suggested that the effects of the co-expressed secretion enhancing factors are partly dependent on the protein to be produced. The microarray based monitoring of the enrichment of genes causing enhanced protein secretory capacity led to novel insights into the limitation of protein secretion.