MilliporeSigma
  • NANOG regulates glioma stem cells and is essential in vivo acting in a cross-functional network with GLI1 and p53.
NANOG regulates glioma stem cells and is essential in vivo acting in a cross-functional network with GLI1 and p53.
The EMBO journal (2010-06-29)
Marie Zbinden, Arnaud Duquet, Aiala Lorente-Trigos, Sandra-Nadia Ngwabyt, Isabel Borges, Ariel Ruiz i Altaba
ABSTRACT
A cohort of genes associated with embryonic stem (ES) cell behaviour, including NANOG, are expressed in a number of human cancers. They form an ES-like signature we first described in glioblastoma multiforme (GBM), a highly invasive and incurable brain tumour. We have also shown that HEDGEHOG-GLI (HH-GLI) signalling is required for GBM growth, stem cell expansion and the expression of this (ES)-like stemness signature. Here, we address the function of NANOG in human GBMs and its relationship with HH-GLI activity. We find that NANOG modulates gliomasphere clonogenicity, CD133(+) stem cell cell behavior and proliferation, and is regulated by HH-GLI signalling. However, GLI1 also requires NANOG activity forming a positive loop, which is negatively controlled by p53 and vice versa. NANOG is essential for GBM tumourigenicity in orthotopic xenografts and it is epistatic to HH-GLI activity. Our data establish NANOG as a novel HH-GLI mediator essential for GBMs. We propose that this function is conserved and that tumour growth and stem cell behaviour rely on the status of a functional GLI1-NANOG-p53 network.