MilliporeSigma
  • Modulation of cardiac vascular endothelial growth factor and PGC-1α with regular postexercise cold-water immersion of rats.
Modulation of cardiac vascular endothelial growth factor and PGC-1α with regular postexercise cold-water immersion of rats.
Journal of applied physiology (Bethesda, Md. : 1985) (2019-01-25)
Ramzi A Al-Horani, Bahaa Al-Trad, Saja Haifawi
ABSTRACT
Myocardial mitochondrial biogenesis and vascular angiogenesis biomarker responses to postexercise cold-water immersion (CWI) have not been reported. Therefore, to determine those cardiac adaptations, adult male Sprague-Dawley rats were divided into three groups: postexercise CWI (CWI; n = 13), exercise only (Ex; n = 12), and untreated control (CON; n = 10). CWI and Ex were trained for 10 wk, 5 sessions/wk, 30-60 min/session. CWI rats were immersed after each session in cold water (15 min at ~12°C). CON remained sedentary. Left ventricle tissue was obtained 48 h after the last exercise session and analyzed for peroxisome proliferator-activated receptor γ coactivator-1α (PGC-1α), vascular endothelial growth factor (VEGF), and heat shock protein 70 kDa (Hsp70) protein content and mRNA expression levels. In addition, superoxide dismutase activity and mRNA and malondialdehyde levels were evaluated. Ex and CWI induced higher PGC-1α protein content compared with CON (1.8 ± 0.6-fold, P < 0.001), which was significantly higher in CWI than Ex rats (P = 0.01). VEGF protein (4.3 ± 3.7-fold) and mRNA (10.1 ± 1.1-fold) were markedly increased only in CWI (P < 0.001) relative to CON. CWI and Ex augmented cardiac Hsp70 protein to a similar level relative to CON (P < 0.05); however, Hsp70 mRNA increased only in Ex (P = 0.002). No further differences were observed between groups. These results suggest that postexercise CWI may further enhance cardiac oxidative capacity by increasing the angiogenic and mitochondrial biogenic factors. In addition, CWI does not seem to worsen exercise-induced cardioprotection and oxidative stress. NEW & NOTEWORTHY A regular postexercise cold-water immersion for 10 wk of endurance training augmented the myocardial mitochondrial biogenesis and vascular angiogenesis coactivators peroxisome proliferator-activated receptor γ coactivator-1α and vascular endothelial growth factor, respectively. In addition, postexercise cold-water immersion did not attenuate the exercise-induced increase in the cardioprotective biomarker heat shock protein 70 kDa or increase exercise-induced oxidative stress.