MilliporeSigma
Slow-Binding Inhibition of Tyrosinase by Ecklonia cava Phlorotannins.
Marine drugs (2019-06-19)
Jang Hoon Kim, Sunggun Lee, Saerom Park, Ji Soo Park, Young Ho Kim, Seo Young Yang
ABSTRACT
Tyrosinase inhibitors improve skin whitening by inhibiting the formation of melanin precursors in the skin. The inhibitory activity of seven phlorotannins (1-7), triphlorethol A (1), eckol (2), 2-phloroeckol (3), phlorofucofuroeckol A (4), 2-O-(2,4,6-trihydroxyphenyl)-6,6'-bieckol (5), 6,8'-bieckol (6), and 8,8'-bieckol (7), from Ecklonia cava was tested against tyrosinase, which converts tyrosine into dihydroxyphenylalanine. Compounds 3 and 5 had IC50 values of 7.0 ± 0.2 and 8.8 ± 0.1 μM, respectively, in competitive mode, with Ki values of 8.2 ± 1.1 and 5.8 ± 0.8 μM. Both compounds showed the characteristics of slow-binding inhibitors over the time course of the enzyme reaction. Compound 3 had a single-step binding mechanism and compound 5 a two-step-binding mechanism. With stable AutoDock scores of -6.59 and -6.68 kcal/mol, respectively, compounds 3 and 5 both interacted with His85 and Asn260 at the active site.
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)