MilliporeSigma
  • MiR-221 is involved in depression by regulating Wnt2/CREB/BDNF axis in hippocampal neurons.
MiR-221 is involved in depression by regulating Wnt2/CREB/BDNF axis in hippocampal neurons.
Cell cycle (Georgetown, Tex.) (2018-12-28)
Nan Lian, Qihui Niu, Yang Lei, Xue Li, Youhui Li, Xueqin Song
ABSTRACT
The aim of this study was to investigate the mechanism of miR-221 in depression. The molecules expressions were measured by qRT-PCR and western blot. The sucrose preference test (SPT), forced swimming test (FST) and tail suspension test (TST) were used to detect depressive-like symptoms. MTT assay and flow cytometric was used to measure the proliferation and apoptosis of hippocampal neuronal. MiR-221 expression in the cerebrospinal fluid and serum of major depressive disorder patients and the hippocampus of chronic unpredictable mild stress (CUMS) mice were increased, while the expression of Wnt2, p-CREB and BDNF were decreased. Additionally, silence of miR-221 increased sucrose preference of CUMS mice and shortened the immobility time of CUMS mice in SPT and FST. MiR-221 could targeted regulate Wnt2, and knockdown of Wnt2 reversed the effect of miR-221 inhibitor on the proliferation and apoptosis of hippocampal neurons and countered the promoting effect of miR-221 inhibitor on the expression of Wnt2, p-CREB and BDNF. MiR-221 could promote the development of depression by regulating Wnt2/CREB/BDNF axis.