MilliporeSigma
  • Intracellular ion and protein nanoparticle-induced osmotic pressure modify astrocyte swelling and brain edema in response to glutamate stimuli.
Intracellular ion and protein nanoparticle-induced osmotic pressure modify astrocyte swelling and brain edema in response to glutamate stimuli.
Redox biology (2019-01-28)
JiaRui Zhang, YuXuan Wang, ZiHui Zheng, XiaoHe Sun, TingTing Chen, Chen Li, XiaoLong Zhang, Jun Guo
ABSTRACT
Intracellular tension activity plays a crucial role in cytotoxic brain edema and astrocyte swelling. Here, a few genetically encoded FRET-based tension probes were designed to detect cytoskeletal structural tension optically, including their magnitude and vectors. The astrocyte swelling resulted in GFAP tension increment, which is associated with the antagonistic effect of inward microfilaments (MFs) and microtubules (MTs) forces. In glutamate-induced astrocyte swelling, GFAP tension rise resulted from outward ion and protein nanoparticle-induced osmotic pressure (PN-OP) increases, where PN-OP could be elicited by MF and MT depolymerization, protein nanoparticle production, and activation of cofilin and stathmin-1. Attenuation of both ion osmotic pressure and PN-OP by drug combinations, together with free-radical scavenger, relieved cerebral edema in vivo. The study suggests that intracellular osmotic pressure (especially PN-OP) has a pivotal role in glutamate-induced astrocyte swelling and brain edema. Recovery of cytoplasmic potential is a promising target to develop new drugs and cure brain edema.