MilliporeSigma
  • MicroRNA-130a regulated by HPV18 E6 promotes proliferation and invasion of cervical cancer cells by targeting TIMP2.
MicroRNA-130a regulated by HPV18 E6 promotes proliferation and invasion of cervical cancer cells by targeting TIMP2.
Experimental and therapeutic medicine (2019-03-25)
Shanlan Yin, Quanle Zhang, Yuhong Wang, Shaoru Li, Ruili Hu
ABSTRACT
Human papillomaviruses (HPVs) have important roles in the development and progression of cervical cancer, but the underlying mechanisms are yet to be fully elucidated. MicroRNA-130a (miR-130a) has previously been reported to promote cervical cancer growth. However, the underlying molecular mechanisms by which miR-130a promotes cervical cancer progression have remained largely elusive. In the present study, polymerase chain reaction and western blot analyses were performed to examine the expression levels of miR-130a and associated proteins. A wound healing assay and a Transwell assay were applied to study cell migration and invasion. A luciferase reporter gene assay was performed to confirm the targeting associations of miR-130a. It was observed that miR-130a was significantly upregulated in cervical cancer tissues compared with that in adjacent non-tumorous tissues. High expression of miR-130a was significantly associated with lymph node metastasis and an advanced clinical stage of cervical cancer. Furthermore, the expression of miR-130a was also higher in HPV(+) cervical cancer cell lines compared with that in HPV(-) cells. Knockdown of HPV18 E6 significantly inhibited the expression of miR-130a in HeLa cervical cancer cells. Furthermore, knockdown of miR-130a reduced the migration and invasion of HeLa cells. Tissue inhibitor of metalloproteinase 2 (TIMP2), an antagonist of matrix metalloproteinase 2 (MMP2), was identified as a novel, direct target gene of miR-130a. The expression of TIMP2 was negatively mediated by miR-130a, and HPV18 E6 inhibited the expression of TIMP2 in HeLa cells. Furthermore, knockdown of TIMP2 rescued the suppressive effects of miR-130a downregulation on the migration and invasion of HeLa cells. In summary, the present study suggests that HPV18 E6 promotes the expression of miR-130a, which further inhibits the expression of TIMP2 and promotes cervical cancer cell invasion. Therefore, HPV/miR-130a/TIMP2 signaling may be a potential target for the prevention of cervical cancer metastasis.