MilliporeSigma
  • Melatonin protects rabbit spermatozoa from cryo-damage via decreasing oxidative stress.
Melatonin protects rabbit spermatozoa from cryo-damage via decreasing oxidative stress.
Cryobiology (2019-04-30)
Zhendong Zhu, Rongnan Li, Yinghua Lv, Wenxian Zeng
ABSTRACT
Mammalian spermatozoa are highly susceptible to reactive oxygen species (ROS) stress. The aim of the present study was to investigate whether and how melatonin protects rabbit spermatozoa against ROS stress during cryopreservation. Semen was diluted with Tris-citrate-glucose extender in presence of different concentrations of melatonin. It was observed that addition of 0.1 mM melatonin significantly improved spermatozoa motility, membrane integrity, acrosome integrity, mitochondrial membrane potential as well as AMP-activated protein kinase (AMPK) phosphorylation. Meanwhile, the lipid peroxidation (LPO), ROS levels and apoptosis of post-thaw spermatozoa were reduced in presence of melatonin. Interestingly, when fresh spermatozoa were incubated with 100 μM H2O2, addition of 0.1 mM melatonin significantly decreased the oxidative damage compared to the H2O2 treatment, whereas addition of luzindole, an MT1 receptor inhibitor, decrease the effect of melatonin in spermatozoa. It was observed that the glutathione (GSH) content and activities of glutathione peroxidase (GPx), superoxide dismutase (SOD) and catalase (CAT) were significantly increased with addition of melatonin during cryopreservation. In conclusion, addition of melatonin to the freezing extender protects rabbit spermatozoa against ROS attack by enhancing AMPK phosphorylation for increasing the antioxidative defense.