MilliporeSigma
  • Accessory heterozygous mutations in cone photoreceptor CNGA3 exacerbate CNG channel-associated retinopathy.
Accessory heterozygous mutations in cone photoreceptor CNGA3 exacerbate CNG channel-associated retinopathy.
The Journal of clinical investigation (2018-11-13)
Markus Burkard, Susanne Kohl, Timm Krätzig, Naoyuki Tanimoto, Christina Brennenstuhl, Anne E Bausch, Katrin Junger, Peggy Reuter, Vithiyanjali Sothilingam, Susanne C Beck, Gesine Huber, Xi-Qin Ding, Anja K Mayer, Britta Baumann, Nicole Weisschuh, Ditta Zobor, Gesa-Astrid Hahn, Ulrich Kellner, Sascha Venturelli, Elvir Becirovic, Peter Charbel Issa, Robert K Koenekoop, Günther Rudolph, John Heckenlively, Paul Sieving, Richard G Weleber, Christian Hamel, Xiangang Zong, Martin Biel, Robert Lukowski, Matthias W Seeliger, Stylianos Michalakis, Bernd Wissinger, Peter Ruth
ABSTRACT
Mutations in CNGA3 and CNGB3, the genes encoding the subunits of the tetrameric cone photoreceptor cyclic nucleotide-gated ion channel, cause achromatopsia, a congenital retinal disorder characterized by loss of cone function. However, a small number of patients carrying the CNGB3/c.1208G>A;p.R403Q mutation present with a variable retinal phenotype ranging from complete and incomplete achromatopsia to moderate cone dysfunction or progressive cone dystrophy. By exploring a large patient cohort and published cases, we identified 16 unrelated individuals who were homozygous or (compound-)heterozygous for the CNGB3/c.1208G>A;p.R403Q mutation. In-depth genetic and clinical analysis revealed a co-occurrence of a mutant CNGA3 allele in a high proportion of these patients (10 of 16), likely contributing to the disease phenotype. To verify these findings, we generated a Cngb3R403Q/R403Q mouse model, which was crossbred with Cnga3-deficient (Cnga3-/-) mice to obtain triallelic Cnga3+/- Cngb3R403Q/R403Q mutants. As in human subjects, there was a striking genotype-phenotype correlation, since the presence of 1 Cnga3-null allele exacerbated the cone dystrophy phenotype in Cngb3R403Q/R403Q mice. These findings strongly suggest a digenic and triallelic inheritance pattern in a subset of patients with achromatopsia/severe cone dystrophy linked to the CNGB3/p.R403Q mutation, with important implications for diagnosis, prognosis, and genetic counseling.
Sigma-Aldrich
Anti-Opsin Antibody, blue, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Opsin Antibody, Red/Green, Chemicon®, from rabbit