MilliporeSigma
  • Cigarette and ENDS preparations differentially regulate ion channels and mucociliary clearance in primary normal human bronchial 3D cultures.
Cigarette and ENDS preparations differentially regulate ion channels and mucociliary clearance in primary normal human bronchial 3D cultures.
American journal of physiology. Lung cellular and molecular physiology (2019-06-06)
Rachael E Rayner, Patrudu Makena, Gaddamanugu L Prasad, Estelle Cormet-Boyaka
ABSTRACT
Cigarette smoking is known to disrupt the normal mucociliary function of the lungs, whereas the effect of electronic nicotine delivery systems (ENDS) is not completely understood. This study aimed to compare the effects of acute exposure of primary normal human bronchial epithelial (NHBE) 3D cultures at air-liquid interface to combustible cigarette and ENDS preparations on mucociliary function, including ion channel function, ciliary beat frequency (CBF), and airway surface liquid (ASL) height. Differentiated NHBE cultures were exposed to whole smoke-conditioned media (WS-CM) or total particulate matter (TPM) prepared from 3R4F reference cigarettes, whole aerosol-conditioned media (ACM) or e-TPM generated from a marketed ENDS product, or nicotine alone. We found that a dose of 7 μg/mL equi-nicotine units of cigarette TPM and WS-CM significantly decreased cystic fibrosis transmembrane conductance regulator (CFTR) and the epithelial sodium channel (ENaC) function, which regulates fluid homeostasis in the lung. Conversely, higher (56 µg/mL) equi-nicotine units of ENDS preparations or nicotine alone had no effect on CFTR and ENaC function. Despite a significant decrease in ion channel function, cigarette smoke preparations did not alter CBF and ASL. Similarly, ENDS preparations and nicotine alone had no effect on ASL and CBF. This study demonstrates that acute exposures of cigarette smoke preparations exert a notable inhibitory effect on CFTR and ENaC function compared with ENDS preparations. In summary, the functional assays described herein are potentially useful for tobacco product evaluations.