MilliporeSigma
  • Targeting Peripheral Somatosensory Neurons to Improve Tactile-Related Phenotypes in ASD Models.
Targeting Peripheral Somatosensory Neurons to Improve Tactile-Related Phenotypes in ASD Models.
Cell (2019-08-10)
Lauren L Orefice, Jacqueline R Mosko, Danielle T Morency, Michael F Wells, Aniqa Tasnim, Shawn M Mozeika, Mengchen Ye, Anda M Chirila, Alan J Emanuel, Genelle Rankin, Ryann M Fame, Maria K Lehtinen, Guoping Feng, David D Ginty
ABSTRACT
Somatosensory over-reactivity is common among patients with autism spectrum disorders (ASDs) and is hypothesized to contribute to core ASD behaviors. However, effective treatments for sensory over-reactivity and ASDs are lacking. We found distinct somatosensory neuron pathophysiological mechanisms underlie tactile abnormalities in different ASD mouse models and contribute to some ASD-related behaviors. Developmental loss of ASD-associated genes Shank3 or Mecp2 in peripheral mechanosensory neurons leads to region-specific brain abnormalities, revealing links between developmental somatosensory over-reactivity and the genesis of aberrant behaviors. Moreover, acute treatment with a peripherally restricted GABAA receptor agonist that acts directly on mechanosensory neurons reduced tactile over-reactivity in six distinct ASD models. Chronic treatment of Mecp2 and Shank3 mutant mice improved body condition, some brain abnormalities, anxiety-like behaviors, and some social impairments but not memory impairments, motor deficits, or overgrooming. Our findings reveal a potential therapeutic strategy targeting peripheral mechanosensory neurons to treat tactile over-reactivity and select ASD-related behaviors.