MilliporeSigma
  • Safety evaluation of 2'-deoxy-2'-fluoro nucleotides in GalNAc-siRNA conjugates.
Safety evaluation of 2'-deoxy-2'-fluoro nucleotides in GalNAc-siRNA conjugates.
Nucleic acids research (2019-03-02)
Maja M Janas, Ivan Zlatev, Ju Liu, Yongfeng Jiang, Scott A Barros, Jessica E Sutherland, Wendell P Davis, Jingxuan Liu, Christopher R Brown, Xiumin Liu, Mark K Schlegel, Lauren Blair, Xuemei Zhang, Biplab Das, Chris Tran, Krishna Aluri, Jing Li, Saket Agarwal, Ramesh Indrakanti, Klaus Charisse, Jayaprakash Nair, Shigeo Matsuda, Kallanthottathil G Rajeev, Tracy Zimmermann, Laura Sepp-Lorenzino, Yuanxin Xu, Akin Akinc, Kevin Fitzgerald, Akshay K Vaishnaw, Peter F Smith, Muthiah Manoharan, Vasant Jadhav, Jing-Tao Wu, Martin A Maier
ABSTRACT
For oligonucleotide therapeutics, chemical modifications of the sugar-phosphate backbone are frequently used to confer drug-like properties. Because 2'-deoxy-2'-fluoro (2'-F) nucleotides are not known to occur naturally, their safety profile was assessed when used in revusiran and ALN-TTRSC02, two short interfering RNAs (siRNAs), of the same sequence but different chemical modification pattern and metabolic stability, conjugated to an N-acetylgalactosamine (GalNAc) ligand for targeted delivery to hepatocytes. Exposure to 2'-F-monomer metabolites was low and transient in rats and humans. In vitro, 2'-F-nucleoside 5'-triphosphates were neither inhibitors nor preferred substrates for human polymerases, and no obligate or non-obligate chain termination was observed. Modest effects on cell viability and mitochondrial DNA were observed in vitro in a subset of cell types at high concentrations of 2'-F-nucleosides, typically not attained in vivo. No apparent functional impact on mitochondria and no significant accumulation of 2'-F-monomers were observed after weekly administration of two GalNAc-siRNA conjugates in rats for ∼2 years. Taken together, the results support the conclusion that 2'-F nucleotides can be safely applied for the design of metabolically stabilized therapeutic GalNAc-siRNAs with favorable potency and prolonged duration of activity allowing for low dose and infrequent dosing.
Sigma-Aldrich
2′,3′-Dideoxycytidine, ≥98% (HPLC)