MilliporeSigma
  • Human gut derived-organoids provide model to study gluten response and effects of microbiota-derived molecules in celiac disease.
Human gut derived-organoids provide model to study gluten response and effects of microbiota-derived molecules in celiac disease.
Scientific reports (2019-05-09)
Rachel Freire, Laura Ingano, Gloria Serena, Murat Cetinbas, Anthony Anselmo, Anna Sapone, Ruslan I Sadreyev, Alessio Fasano, Stefania Senger
ABSTRACT
Celiac disease (CD) is an immune-mediated disorder triggered by gluten exposure. The contribution of the adaptive immune response to CD pathogenesis has been extensively studied, but the absence of valid experimental models has hampered our understanding of the early steps leading to loss of gluten tolerance. Using intestinal organoids developed from duodenal biopsies from both non-celiac (NC) and celiac (CD) patients, we explored the contribution of gut epithelium to CD pathogenesis and the role of microbiota-derived molecules in modulating the epithelium's response to gluten. When compared to NC, RNA sequencing of CD organoids revealed significantly altered expression of genes associated with gut barrier, innate immune response, and stem cell functions. Monolayers derived from CD organoids exposed to gliadin showed increased intestinal permeability and enhanced secretion of pro-inflammatory cytokines compared to NC controls. Microbiota-derived bioproducts butyrate, lactate, and polysaccharide A improved barrier function and reduced gliadin-induced cytokine secretion. We concluded that: (1) patient-derived organoids faithfully express established and newly identified molecular signatures characteristic of CD. (2) microbiota-derived bioproducts can be used to modulate the epithelial response to gluten. Finally, we validated the use of patient-derived organoids monolayers as a novel tool for the study of CD.
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)