MilliporeSigma
  • Brain-derived neurotrophic factor promotes tumorigenesis via induction of neovascularization: implication in hepatocellular carcinoma.
Brain-derived neurotrophic factor promotes tumorigenesis via induction of neovascularization: implication in hepatocellular carcinoma.
Clinical cancer research : an official journal of the American Association for Cancer Research (2011-03-23)
Chi-Tat Lam, Zhen-Fan Yang, Chi-Keung Lau, Ka-Ho Tam, Sheung-Tat Fan, Ronnie T P Poon
ABSTRACT
Brain-derived neurotrophic factor (BDNF) has emerged as a novel angiogenic factor, and yet its impact on tumorigenesis is unclear. This study aimed at investigating the roles of BDNF in angiogenesis and tumor development. BDNF was overexpressed in a mouse endothelial cell (EC) line by stable transfection, and angiogenic properties of the transfectants were assessed. Microarray analysis was employed to explore the molecular pathways. The impact of modulating BDNF levels in two mouse EC lines on tumorigenic potential of a transformed mouse liver cell line was evaluated by an in vivo cotransplantation model. BDNF and tropomyosin receptor kinase B (TrkB) protein levels were determined in 50 pairs of human hepatocellular carcinoma (HCC) tissues by Western blotting and immunohistochemistry. Survival analysis was carried out to determine their clinical significance. Overexpression of BDNF could promote EC proliferation, migration, invasion, and survival. Microarray and molecular studies showed that RhoA, caspase-9, caspase-3, growth arrest specific 6, and VEGF could mediate BDNF/TrkB-induced angiogenesis. The cotransplantation experiment showed that high BDNF-expressing ECs could facilitate tumor angiogenesis and growth, whereas knockdown of BDNF by short hairpin RNAs impaired such effects. Furthermore, examination on human HCC tissues revealed upregulation of BDNF and TrkB protein levels in 46.0% and 33.3% of the cases studied, respectively. Immunohistochemistry disclosed strong BDNF reactivity in both tumor and endothelial cells. High TrkB expression was associated with shorter overall survival. BDNF/TrkB system was crucial for tumor angiogenesis and growth, which may represent a potential target for antiangiogenic therapy in HCC.
Sigma-Aldrich
Blood Vessel Staining Kit, Reagents & materials supplied in the Blood Vessel Staining Kit are sufficient for staining 100 tissue sections & when stored at 2-8ºC is stable up to the expiration date.