MilliporeSigma
  • Protection against cisplatin-induced ovarian damage by the antioxidant sodium 2-mercaptoethanesulfonate (mesna) in female rats.
Protection against cisplatin-induced ovarian damage by the antioxidant sodium 2-mercaptoethanesulfonate (mesna) in female rats.
American journal of obstetrics and gynecology (2008-04-09)
John Yeh, Beom Su Kim, Jennifer Peresie
ABSTRACT
The hypothesis was that the administration of the antioxidant mesna (sodium 2-mercaptoethanesulfonate) during chemotherapy would protect ovaries against follicular damage. Sprague-Dawley rats were treated with saline solution, mesna-plus-cisplatin, or cisplatin. Immunohistochemistry was used to evaluate the Müllerian inhibiting substance (MIS) positive follicles. Serum and ovarian MIS were measured with enzyme-linked immunosorbent assay and Western blot analysis, respectively. Apoptosis in ovaries was studied by terminal deoxynucleotidyl transfer biotin-d UTP nick end labeling (TUNEL) assay. Immunofluorescence staining for MIS was higher in preantral follicles in the mesna-plus-cisplatin group. The ovarian and serum MIS levels were higher in the mesna-plus-cisplatin than in the cisplatin alone group. There were no differences statistically in the TUNEL and the ovarian cyst analyses. Mesna, which was used at the time of cisplatin administration, protected ovaries against damage. The data that are presented challenge the existing clinical paradigm that gonadotropin-releasing hormone agonists represent the only medical method for the protection of ovaries during chemotherapy. Alternative medical means to protect ovaries during chemotherapy may be achievable.
Sigma-Aldrich
ApopTag Plus In Situ Apoptosis Fluorescein Detection Kit, The ApopTag Plus Fluorescein In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the indirect TUNEL method, utilizing an anti-digoxigenin antibody that is conjugated to a fluorescein reporter molecule.