MilliporeSigma
Identification of a major co-receptor for primary isolates of HIV-1.
Nature (1996-06-20)
H Deng, R Liu, W Ellmeier, S Choe, D Unutmaz, M Burkhart, P Di Marzio, S Marmon, R E Sutton, C M Hill, C B Davis, S C Peiper, T J Schall, D R Littman, N R Landau
ABSTRACT
Entry of HIV-1 into target cells requires cell-surface CD4 and additional host cell cofactors. A cofactor required for infection with virus adapted for growth in transformed T-cell lines was recently identified and named fusin. However, fusin does not promote entry of macrophage-tropic viruses, which are believed to be the key pathogenic strains in vivo. The principal cofactor for entry mediated by the envelope glycoproteins of primary macrophage-tropic strains of HIV-1 is CC-CKR-5, a receptor for the beta-chemokines RANTES, MIP-1alpha and MIP-1beta.